Assay Details
| Gene Symbol: Myb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198914.1 | NP_001185843.1 | 13 - 14 | 2201 | 78 | |
| NM_010848.3 | NP_034978.3 | 12 - 13 | 1844 | 78 | ||
| GenBank mRNA | AK088079.1 | - | 13 - 14 | 2122 | 78 | |
| AK132250.1 | - | 12 - 13 | 1709 | 78 | ||
| AK137965.1 | - | 12 - 13 | 1710 | 78 | ||
| BC011513.1 | - | 12 - 13 | 1752 | 3672769 | 78 | |
| M12848.1 | - | 12 - 13 | 1842 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myb
Genomic Map