Assay Details
| Gene Symbol: Fyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC032149.1 | - | 1 - 2 | 81 | 3602546 | 64 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC032149.1 transcripts.
Set Membership: |
|

Fyn
Genomic Map