Assay Details
| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB041555.1 | - | 4 - 5 | 912 | 67 | |
| AK169598.1 | - | 4 - 5 | 936 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsrp1| Gene Symbol: Rsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB041555.1 | - | 4 - 5 | 912 | 67 | |
| AK169598.1 | - | 4 - 5 | 936 | 67 | ||
Set Membership: |
|