Assay Details
| Gene Symbol: Rragc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017475.2 | NP_059503.2 | 4 - 5 | 851 | 82 | |
| GenBank mRNA | AB017616.1 | - | 4 - 5 | 774 | 82 | |
| AK159355.1 | - | 4 - 5 | 784 | 82 | ||
| AK159434.1 | - | 4 - 5 | 806 | 82 | ||
| AK166658.1 | - | 4 - 5 | 797 | 82 | ||
| BC005417.1 | - | 4 - 5 | 799 | 3495022 | 82 | |
| BC037732.1 | - | 1 - 2 | 148 | 82 | ||
| BC071245.1 | - | 4 - 5 | 1199 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rragc
Genomic Map