Assay Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290688.1 | NP_001277617.1 | 4 - 5 | 410 | 115 | |
| NM_020000.3 | NP_064384.2 | 4 - 5 | 450 | 115 | ||
| NM_173719.3 | NP_776067.1 | 5 - 6 | 498 | 115 | ||
| GenBank mRNA | AB041805.1 | - | 3 - 4 | 1936 | 115 | |
| AK005422.1 | - | 5 - 6 | 481 | 115 | ||
| AK008759.1 | - | 4 - 5 | 429 | 115 | ||
| AK011080.1 | - | 4 - 5 | 410 | 115 | ||
| AK147032.1 | - | 4 - 5 | 436 | 115 | ||
| AK148025.1 | - | 4 - 5 | 431 | 115 | ||
| BC002315.1 | - | 3 - 4 | 390 | 115 | ||
| BC021870.1 | - | 4 - 5 | 421 | 4022630 | 115 | |
| BC051683.1 | - | 4 - 5 | 388 | 115 | ||
| BC053738.1 | - | 2 - 3 | 1297 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med8
Genomic Map