Assay Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173719.3 | NP_776067.1 | 2 - 3 | 170 | 105 | |
| GenBank mRNA | AK005422.1 | - | 2 - 3 | 153 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med8
Genomic Map