Assay Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290688.1 | NP_001277617.1 | 2 - 3 | 165 | 118 | |
| GenBank mRNA | AK011080.1 | - | 2 - 3 | 165 | 118 | |
| BC051683.1 | - | 2 - 3 | 143 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011080.1, NM_001290688.1 transcripts.
Set Membership: |
|

Med8
Genomic Map