Assay Details
| Gene Symbol: Stk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009288.2 | NP_033314.2 | 9 - 10 | 1691 | 69 | |
| GenBank mRNA | AK148118.1 | - | 9 - 10 | 1634 | 69 | |
| AK220448.1 | - | 9 - 10 | 1613 | 69 | ||
| BC069840.1 | - | 9 - 10 | 1764 | 69 | ||
| BC096624.1 | - | 9 - 10 | 1776 | 69 | ||
| BC128363.1 | - | 9 - 10 | 1577 | 40111177 | 69 | |
| D89728.1 | - | 9 - 10 | 1652 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk10
Genomic Map