Assay Details
| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027930.3 | NP_082206.2 | 4 - 5 | 427 | 115 | |
| GenBank mRNA | AK011410.1 | - | 4 - 5 | 422 | 115 | |
| AK016786.1 | - | 4 - 5 | 472 | 115 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtfr2
Genomic Map