Assay Details
| Gene Symbol: Mtfr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256112.1 | NP_001243041.1 | 4 - 5 | 525 | 64 | |
| NM_029759.4 | NP_084035.1 | 4 - 5 | 476 | 64 | ||
| GenBank mRNA | AK010825.1 | - | 4 - 5 | 517 | 64 | |
| AK080126.1 | - | 3 - 4 | 2845 | 64 | ||
| AK089141.1 | - | 4 - 5 | 517 | 64 | ||
| AK145362.1 | - | 4 - 5 | 517 | 64 | ||
| AK146416.1 | - | 3 - 4 | 352 | 64 | ||
| AK164004.1 | - | 3 - 4 | 352 | 64 | ||
| AK169047.1 | - | 4 - 5 | 384 | 64 | ||
| AK171703.1 | - | 4 - 5 | 517 | 64 | ||
| AK187934.1 | - | 4 - 5 | 357 | 64 | ||
| BC026808.1 | - | 3 - 4 | 348 | 4159289 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtfr1l
Genomic Map