Assay Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030189.3 | NP_084465.1 | 5 - 6 | 690 | 69 | |
| GenBank mRNA | AK015061.1 | - | 6 - 7 | 884 | 69 | |
| AK019766.1 | - | 5 - 6 | 691 | 69 | ||
| BC145657.1 | - | 4 - 5 | 477 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stpg1
Genomic Map