| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400028645.mm.1 |
TC0400003529.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028645.mm.1 |
NM_001162533.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028645.mm.1 |
NM_001162533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028645.mm.1 |
NM_025856.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028645.mm.1 |
NM_025856.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028650.mm.1 |
TC0400003529.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028650.mm.1 |
NM_001162533.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028650.mm.1 |
NM_001162533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028652.mm.1 |
TC0400003529.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028652.mm.1 |
NM_001162533.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028652.mm.1 |
NM_001162533.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028651.mm.1 |
TC0400003529.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028651.mm.1 |
NM_001162533.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028651.mm.1 |
NM_001162533.1 |
Refseq |