Assay Details
| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145970.1 | NP_001139442.1 | 12 - 13 | 2188 | 72 | |
| NM_144941.3 | NP_659190.3 | 12 - 13 | 2404 | 72 | ||
| GenBank mRNA | AK122462.1 | - | 6 - 7 | 3827 | 72 | |
| AK151613.1 | - | 10 - 11 | 1919 | 72 | ||
| BC016081.1 | - | 3 - 4 | 440 | 72 | ||
| BC019977.1 | - | 7 - 8 | 1380 | 4187506 | 72 | |
| BC023677.1 | - | 12 - 13 | 2379 | 5354497 | 72 | |
| BC023774.1 | - | 12 - 13 | 2185 | 72 | ||
| BM944422.1 | - | 3 - 4 | 294 | 72 | ||
More Information
Set Membership: |
|

Map7d1
Genomic Map