| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400028719.mm.1 |
TC0400003533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028719.mm.1 |
NM_001145970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028719.mm.1 |
NM_001145970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028719.mm.1 |
NM_144941.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028719.mm.1 |
NM_144941.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028721.mm.1 |
TC0400003533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028721.mm.1 |
NM_001145970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028721.mm.1 |
NM_001145970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028721.mm.1 |
NM_144941.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028721.mm.1 |
NM_144941.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028722.mm.1 |
TC0400003533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028722.mm.1 |
NM_001145970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028722.mm.1 |
NM_001145970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028722.mm.1 |
NM_144941.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028722.mm.1 |
NM_144941.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028718.mm.1 |
TC0400003533.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400028718.mm.1 |
NM_001145970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028718.mm.1 |
NM_001145970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028718.mm.1 |
NM_144941.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400028718.mm.1 |
NM_144941.3 |
Refseq |