Assay Details
| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145970.1 | NP_001139442.1 | 2 - 3 | 649 | 63 | |
| NM_144941.3 | NP_659190.3 | 2 - 3 | 649 | 63 | ||
| GenBank mRNA | AK151613.1 | - | 2 - 3 | 637 | 63 | |
| CV563343.1 | - | 1 - 2 | 220 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map7d1
Genomic Map