Assay Details
| Gene Symbol: Sh3d21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025856.3 | NP_080132.1 | 2 - 3 | 264 | 89 | |
| GenBank mRNA | AK076962.1 | - | 2 - 3 | 229 | 89 | |
| BC025637.1 | - | 2 - 3 | 275 | 89 | ||
| BC053421.1 | - | 2 - 3 | 225 | 6706860 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076962.1 transcripts.
Set Membership: |
|

Sh3d21
Genomic Map