Assay Details
| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153177.3 | NP_694817.2 | 1 - 2 | 276 | 61 | |
| GenBank mRNA | AK030018.1 | - | 1 - 2 | 292 | 61 | |
| BB616446.1 | - | 1 - 2 | 276 | 61 | ||
| BC096023.1 | - | 1 - 2 | 58 | 30607807 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC096023.1, NM_153177.3 transcripts.
Set Membership: |
|

Ago4
Genomic Map