Assay Details
| Gene Symbol: Zxdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173002.3 | NP_766590.1 | 9 - 10 | 2662 | 68 | |
| GenBank mRNA | AK044429.1 | - | 9 - 10 | 2555 | 68 | |
| AK047079.1 | - | 9 - 10 | 2544 | 68 | ||
| AK047538.1 | - | 9 - 10 | 2578 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zxdc
Genomic Map