Assay Details
| Gene Symbol: Trim63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039048.2 | NP_001034137.2 | 3 - 4 | 605 | 67 | |
| GenBank mRNA | BC147663.1 | - | 3 - 4 | 573 | 9087452 | 67 |
| BC147666.1 | - | 3 - 4 | 573 | 9087455 | 67 | |
| DQ229108.1 | - | 3 - 4 | 501 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
regulation of gene expression
response to electrical stimulus involved in regulation of muscle adaptation
response to denervation involved in regulation of muscle adaptation
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
cellular response to dexamethasone stimulus
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
titin binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
muscle contraction
regulation of gene expression
response to electrical stimulus involved in regulation of muscle adaptation
response to denervation involved in regulation of muscle adaptation
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
cellular response to dexamethasone stimulus
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
titin binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106745 | DQ229108.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106745 | BC147663.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106745 | BC147666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106745 | CK128385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106745 | NM_001039048.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159109 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140768 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168709 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165631 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106745 | DQ229108.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106745 | BC147663.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106745 | BC147666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106745 | CK128385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106745 | NM_001039048.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P115335 | AK052911.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168709 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140768 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00994571 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00994578 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165631 | - | - |
| Whole Mouse Genome G4122A | A_51_P115335 | AK052911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P517224 | NM_001039048.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P517224 | AJ278734.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140768 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168709 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03681170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03629171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03282985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03761014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03312776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03761010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03538271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03730060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03355175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03346691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03538269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03681166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03602166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03761011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03616246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03602179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03547322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03696881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03346693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03542171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03761008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03879942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03648882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322155 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10508954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508956 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438704_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10508953 | - | - |
| Mouse Genome 430 2.0 Array | 1438704_at | - | - |
| Mouse Genome 430 2.0 Array | 1438704_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433766_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 433766_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400012968.mm.1 | NM_001039048.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012968.mm.1 | NM_001039048.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012968.mm.1 | TC0400001631.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438704_PM_at | - | - |
| Mouse Expression Set 430 | 1438704_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438704_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438704_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106290170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106290170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161061 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161061 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215654 | - | - |
| mouse-6 v1.1 (Illumina) | GI_84794632-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106290170 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106290170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161060 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215654 | - | - |

Trim63
Genomic Map