Assay Details
| Gene Symbol: Snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042451.2 | NP_001035916.1 | 1 - 2 | 283 | 108 | |
| GenBank mRNA | AF033261.1 | - | 1 - 2 | 110 | 108 | |
| AK014472.1 | - | 1 - 2 | 152 | 108 | ||
| AK156316.1 | - | 1 - 2 | 119 | 108 | ||
| BY099071.1 | - | 1 - 2 | 275 | 108 | ||
| BY782503.1 | - | 1 - 2 | 284 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF033261.1, AK014472.1, AK156316.1, NM_001042451.2 transcripts.
Set Membership: |
|

Snca
Genomic Map