Assay Details
| Gene Symbol: Snca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009221.2 | NP_033247.1 | 1 - 2 | 188 | 88 | |
| GenBank mRNA | AF179273.1 | - | 1 - 2 | 188 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF179273.1, NM_009221.2 transcripts.
Set Membership: |
|

Snca
Genomic Map