Assay Details
| Gene Symbol: Rad52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166381.1 | NP_001159853.1 | 7 - 8 | 694 | 136 | |
| NM_001166382.1 | NP_001159854.1 | 7 - 8 | 694 | 136 | ||
| NM_001166383.1 | NP_001159855.1 | 7 - 8 | 694 | 136 | ||
| NM_011236.2 | NP_035366.2 | 7 - 8 | 694 | 136 | ||
| GenBank mRNA | AK080702.1 | - | 7 - 8 | 667 | 136 | |
| BC010673.1 | - | 6 - 7 | 566 | 136 | ||
| BC019123.1 | - | 7 - 8 | 674 | 5003696 | 136 | |
| U06837.1 | - | 7 - 8 | 616 | 136 | ||
| U12135.1 | - | 7 - 8 | 663 | 136 | ||
| Z32767.1 | - | 7 - 8 | 688 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad52
Genomic Map