Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104484.1 | 19 - 20 | 3535 | 113 | ||
| GenBank mRNA | AF340028.1 | - | 19 - 20 | 3519 | 113 | |
| AK122437.1 | - | 8 - 9 | 898 | 113 | ||
More Information
Set Membership: |
|

Erc1
Genomic Map