Assay Details
| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011909.2 | NP_036039.2 | 6 - 7 | 883 | 84 | |
| GenBank mRNA | AF069502.1 | - | 6 - 7 | 844 | 84 | |
| AK149772.1 | - | 6 - 7 | 884 | 84 | ||
| AK151011.1 | - | 6 - 7 | 884 | 84 | ||
| AK151260.1 | - | 6 - 7 | 884 | 84 | ||
| AK151722.1 | - | 6 - 7 | 884 | 84 | ||
| AK152034.1 | - | 6 - 7 | 849 | 84 | ||
| AK152574.1 | - | 6 - 7 | 849 | 84 | ||
| AK153312.1 | - | 6 - 7 | 853 | 84 | ||
| BC138562.1 | - | 6 - 7 | 765 | 8861582 | 84 | |
| BC138577.1 | - | 6 - 7 | 765 | 8861597 | 84 | |
| CT010228.1 | - | 5 - 6 | 619 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp18
Genomic Map