Assay Details
| Gene Symbol: C1rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181344.5 | NP_851989.3 | 1 - 2 | 119 | 59 | |
| GenBank mRNA | AK038659.1 | - | 1 - 2 | 84 | 59 | |
| AK133495.1 | - | 1 - 2 | 98 | 59 | ||
| AK134207.1 | - | 1 - 2 | 120 | 59 | ||
| AK162497.1 | - | 1 - 2 | 74 | 59 | ||
| BC070472.1 | - | 1 - 2 | 82 | 30627947 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070472.1 transcripts.
Set Membership: |
|

C1rl
Genomic Map