Assay Details
| Gene Symbol: Ccdc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034876.1 | NP_001030048.1 | 3 - 4 | 386 | 60 | |
| GenBank mRNA | AK133118.1 | - | 3 - 4 | 387 | 60 | |
| BG088507.2 | - | 3 - 4 | 311 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc24
Genomic Map