Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 11 - 12 | 1271 | 92 | |
| GenBank mRNA | AK090104.1 | - | 10 - 11 | 1095 | 92 | |
| AK144547.1 | - | 11 - 12 | 1272 | 92 | ||
| AK165019.1 | - | 11 - 12 | 1215 | 92 | ||
| BC059244.1 | - | 2 - 3 | 182 | 92 | ||
| BC144781.1 | - | 11 - 12 | 1214 | 9053329 | 92 | |
| BC145693.1 | - | 11 - 12 | 1214 | 40130996 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map