Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 13 - 14 | 1439 | 85 | |
| GenBank mRNA | AK090104.1 | - | 12 - 13 | 1263 | 85 | |
| AK144547.1 | - | 13 - 14 | 1440 | 85 | ||
| AK165019.1 | - | 12 - 13 | 2059 | 85 | ||
| BC059244.1 | - | 4 - 5 | 350 | 85 | ||
| BC144781.1 | - | 13 - 14 | 1382 | 9053329 | 85 | |
| BC145693.1 | - | 13 - 14 | 1382 | 40130996 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map