Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 19 - 20 | 2228 | 79 | |
| GenBank mRNA | AK016259.1 | - | 3 - 4 | 333 | 79 | |
| AK220360.1 | - | 4 - 5 | 502 | 79 | ||
| BC043037.1 | - | 2 - 3 | 319 | 79 | ||
| BC059244.1 | - | 10 - 11 | 1139 | 79 | ||
| BC144781.1 | - | 19 - 20 | 2171 | 9053329 | 79 | |
| BC145693.1 | - | 19 - 20 | 2171 | 40130996 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map