Assay Details
| Gene Symbol: Klrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099918.1 | NP_001093388.1 | 4 - 5 | 406 | 73 | |
| GenBank mRNA | DQ113419.1 | - | 4 - 5 | 406 | 73 | |
| HQ713436.1 | - | 4 - 5 | 406 | 73 | ||
| JF429832.1 | - | 4 - 5 | 419 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968890 | AK015258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077956 | DQ113419.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077956 | NM_001099918.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 5483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | AK004652.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | AK082323.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | AK082761.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | AK141692.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | AK145969.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | BC034517.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | NM_007949.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584670 | U97572.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 9817 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077956 | DQ113419.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077956 | NM_001099918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968890 | AK015258.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24840438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05486136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05215895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05201513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05333441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05784915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05222418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05873186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05545534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05769004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05873183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05201512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05784902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05156159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05784900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05873184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05784914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24840381 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548306 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548301 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548300 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025121.mm.1 | TC0600003213.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025121.mm.1 | NM_001099918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025121.mm.1 | NM_001099918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025122.mm.1 | TC0600003213.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025122.mm.1 | NM_001099918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025122.mm.1 | NM_001099918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025125.mm.1 | TC0600003213.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025125.mm.1 | NM_001099918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025125.mm.1 | NM_001099918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025124.mm.1 | TC0600003213.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025124.mm.1 | NM_001099918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025124.mm.1 | NM_001099918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025123.mm.1 | TC0600003213.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025123.mm.1 | NM_001099918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025123.mm.1 | NM_001099918.1 | Refseq |
| Murine Genome U74v2 Set | 170600_PM_at | - | - |
| Murine Genome U74v2 Set | 170600_at | - | - |

Klrb1
Genomic Map