Assay Details
| Gene Symbol: Csgalnact2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030165.3 | NP_084441.3 | 1 - 2 | 118 | 169 | |
| GenBank mRNA | AK019496.1 | - | 1 - 2 | 121 | 169 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019496.1, NM_030165.3 transcripts.
Set Membership: |
|

Csgalnact2
Genomic Map