Assay Details
| Gene Symbol: Ctnna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109764.1 | NP_001103234.1 | 14 - 15 | 2245 | 78 | |
| NM_009819.2 | NP_033949.2 | 13 - 14 | 2143 | 78 | ||
| NM_145732.2 | NP_663785.2 | 14 - 15 | 2245 | 78 | ||
| GenBank mRNA | AK158916.1 | - | 14 - 15 | 2278 | 78 | |
| BC051995.1 | - | 13 - 14 | 2024 | 78 | ||
| BC079648.1 | - | 14 - 15 | 2162 | 6850068 | 78 | |
| D25281.1 | - | 14 - 15 | 2172 | 78 | ||
| D25282.1 | - | 13 - 14 | 2072 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnna2
Genomic Map