Assay Details
| Gene Symbol: Ctnna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109764.1 | NP_001103234.1 | 1 - 2 | 249 | 73 | |
| NM_145732.2 | NP_663785.2 | 1 - 2 | 249 | 73 | ||
| GenBank mRNA | AK158916.1 | - | 1 - 2 | 282 | 73 | |
| BC079648.1 | - | 1 - 2 | 166 | 6850068 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158916.1, BC079648.1, NM_001109764.1, NM_145732.2 transcripts.
Set Membership: |
|

Ctnna2
Genomic Map