Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
TC0600002519.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_001109764.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_001109764.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_009819.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_009819.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_145732.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019559.mm.1 |
NM_145732.2 |
Refseq |