Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177884.3 | NP_808552.2 | 13 - 14 | 1894 | 162 | |
| GenBank mRNA | AK038369.1 | - | 14 - 15 | 1881 | 162 | |
| AK050789.1 | - | 13 - 14 | 1836 | 162 | ||
| AK078171.1 | - | 13 - 14 | 1884 | 162 | ||
| AK136387.1 | - | 11 - 12 | 1496 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcfc2
Genomic Map