Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177884.3 | NP_808552.2 | 4 - 5 | 718 | 62 | |
| GenBank mRNA | AK050789.1 | - | 4 - 5 | 660 | 62 | |
| AK077263.1 | - | 4 - 5 | 708 | 62 | ||
| AK078171.1 | - | 4 - 5 | 709 | 62 | ||
| AK136387.1 | - | 2 - 3 | 320 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcfc2
Genomic Map