Assay Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177884.3 | NP_808552.2 | 9 - 10 | 131 | ||
| GenBank mRNA | AK050789.1 | - | 9 - 10 | 131 | ||
| AK078171.1 | - | 9 - 10 | 131 | |||
| AK136387.1 | - | 7 - 8 | 131 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gcfc2
Genomic Map