Assay Details
| Gene Symbol: Yars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134151.4 | NP_598912.4 | 4 - 5 | 732 | 73 | |
| GenBank mRNA | AK028785.1 | - | 4 - 5 | 594 | 73 | |
| AK043837.1 | - | 4 - 5 | 656 | 73 | ||
| AK076634.1 | - | 4 - 5 | 577 | 73 | ||
| AK139579.1 | - | 4 - 5 | 577 | 73 | ||
| AK145356.1 | - | 4 - 5 | 625 | 73 | ||
| AK151880.1 | - | 4 - 5 | 584 | 73 | ||
| AK169708.1 | - | 4 - 5 | 508 | 73 | ||
| BC013552.1 | - | 4 - 5 | 663 | 4236514 | 73 | |
| BC022143.1 | - | 4 - 5 | 627 | 5101324 | 73 | |
| BC026615.1 | - | 4 - 5 | 639 | 4976999 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yars
Genomic Map