Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000014145.mm.1 |
NM_001101452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014145.mm.1 |
NM_001101452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014145.mm.1 |
TC1000001986.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014147.mm.1 |
NM_001101452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014147.mm.1 |
NM_001101452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014147.mm.1 |
TC1000001986.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000014146.mm.1 |
NM_001101452.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014146.mm.1 |
NM_001101452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000014146.mm.1 |
TC1000001986.mm.1 |
MTA TC |