Assay Details
| Gene Symbol: Gm4794 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101452.1 | NP_001094922.1 | 3 - 4 | 481 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723747 | AK044004.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512822 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735322 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30787 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P301602 | NM_001101452.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27053760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08918916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27053778 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368607 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368608 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368606 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000014143.mm.1 | NM_001101452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014143.mm.1 | NM_001101452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014143.mm.1 | TC1000001986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014142.mm.1 | NM_001101452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014142.mm.1 | NM_001101452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014142.mm.1 | TC1000001986.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000014144.mm.1 | NM_001101452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014144.mm.1 | NM_001101452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000014144.mm.1 | TC1000001986.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 682747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226400 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226400 | - | - |

Gm4794
Genomic Map