Assay Details
| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028287.2 | NP_082563.1 | 4 - 5 | 883 | 94 | |
| GenBank mRNA | AK046191.1 | - | 4 - 5 | 886 | 94 | |
| AK052071.1 | - | 4 - 5 | 867 | 94 | ||
| AK077304.1 | - | 4 - 5 | 931 | 94 | ||
| AK077730.1 | - | 4 - 5 | 904 | 94 | ||
| AK160928.1 | - | 4 - 5 | 882 | 94 | ||
| AK164710.1 | - | 2 - 3 | 2398 | 94 | ||
| BC067065.1 | - | 3 - 4 | 797 | 94 | ||
| BC089514.1 | - | 4 - 5 | 896 | 30252639 | 94 | |
More Information
Set Membership: |
|

Zufsp
Genomic Map