Assay Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145148.2 | NP_660130.2 | 2 - 3 | 584 | 76 | |
| GenBank mRNA | AK051779.1 | - | 2 - 3 | 456 | 76 | |
| AK132280.1 | - | 2 - 3 | 448 | 76 | ||
| BC052390.1 | - | 2 - 3 | 453 | 5718942 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132280.1, BC052390.1, NM_145148.2 transcripts.
Set Membership: |
|

Frmd4b
Genomic Map