Assay Details
| Gene Symbol: Wasf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031877.3 | NP_114083.1 | 4 - 5 | 523 | 129 | |
| GenBank mRNA | AF290877.1 | - | 3 - 4 | 419 | 129 | |
| AF467773.1 | - | 3 - 4 | 419 | 129 | ||
| BC016896.1 | - | 4 - 5 | 481 | 4239038 | 129 | |
| BC053423.1 | - | 4 - 5 | 484 | 6509953 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wasf1
Genomic Map