Assay Details
| Gene Symbol: Wasf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031877.3 | NP_114083.1 | 6 - 7 | 823 | 68 | |
| GenBank mRNA | AF290877.1 | - | 5 - 6 | 719 | 68 | |
| AF467773.1 | - | 5 - 6 | 719 | 68 | ||
| BC016896.1 | - | 6 - 7 | 781 | 4239038 | 68 | |
| BC053423.1 | - | 6 - 7 | 784 | 6509953 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wasf1
Genomic Map