Assay Details
| Gene Symbol: Zfa-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037920.1 | 1 | 388 | 90 | ||
| GenBank mRNA | AK029509.1 | - | 1 | 389 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029509.1 transcripts.
Set Membership: |
|

Zfa-ps
Genomic Map