Assay Details
| Gene Symbol: Eogt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175313.4 | NP_780522.1 | 15 - 16 | 1716 | 69 | |
| GenBank mRNA | AK037507.1 | - | 15 - 16 | 1717 | 69 | |
| AK201418.1 | - | 4 - 5 | 311 | 69 | ||
| AK205004.1 | - | 5 - 6 | 321 | 69 | ||
| BC048939.1 | - | 13 - 14 | 1255 | 6439908 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eogt
Genomic Map