Assay Details
| Gene Symbol: Tead4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080979.1 | NP_001074448.2 | 8 - 9 | 1312 | 64 | |
| NM_011567.2 | NP_035697.3 | 9 - 10 | 1441 | 64 | ||
| GenBank mRNA | AB221623.1 | - | 7 - 8 | 742 | 64 | |
| AK133915.1 | - | 9 - 10 | 1103 | 64 | ||
| BC130257.1 | - | 7 - 8 | 747 | 40142595 | 64 | |
| D87965.1 | - | 8 - 9 | 1312 | 64 | ||
| D87966.1 | - | 9 - 10 | 1436 | 64 | ||
| L26343.1 | - | 9 - 10 | 981 | 64 | ||
| L26344.1 | - | 926 | 64 | |||
| U51743.1 | - | 7 - 8 | 727 | 64 | ||
| X94441.1 | - | 8 - 9 | 872 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead4
Genomic Map