Assay Details
| Gene Symbol: Tead4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011567.2 | NP_035697.3 | 5 - 6 | 1036 | 85 | |
| GenBank mRNA | AK133915.1 | - | 5 - 6 | 697 | 85 | |
| D87966.1 | - | 5 - 6 | 1031 | 85 | ||
| X94441.1 | - | 4 - 5 | 467 | 85 | ||
More Information
Set Membership: |
|

Tead4
Genomic Map