Assay Details
| Gene Symbol: Cracr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033464.3 | NP_001028636.1 | 2 - 3 | 355 | 92 | |
| GenBank mRNA | AK143832.1 | - | 2 - 3 | 356 | 92 | |
| BC150869.1 | - | 2 - 3 | 433 | 9087780 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cracr2a
Genomic Map