Assay Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028604.2 | NP_082880.2 | 8 - 9 | 848 | 78 | |
| GenBank mRNA | AF532979.1 | - | 8 - 9 | 848 | 78 | |
| AK010722.1 | - | 8 - 9 | 790 | 78 | ||
| BC048703.1 | - | 6 - 7 | 523 | 6509593 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt11
Genomic Map